Learn More
Sino Biological Recombinant Human Apolipoprotein A-I/APOA1 Protein (hFc Tag)
Supplier:  Sino Biological 10686H02H100
Encompass_Preferred
A DNA sequence encoding the pro-form of human APOA1 (CAA26097.1) (Met 1-Gln 267) was fused with Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5389