Learn More
Sino Biological Recombinant Mouse IgG1-Fc Protein (C102S), HPLC-verified
Supplier:  Sino Biological 10690MNAH100
Encompass_Preferred
The Fc region of mouse IgG1 (P01868-1) (Val 98-Lys 324) (one aa mutation, 102 Cys / Ser) was expressed and purified.
Catalog No. 50-161-5403