Learn More
Sino Biological Recombinant Human Angiopoietin-2/ANGPT2 Protein (His Tag)
Supplier:  Sino Biological 10691H08H50
Encompass_Preferred
A DNA sequence encoding the human Angiopoietin 2 (NP_001138.1) (Met 1-Phe 496) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5412