Learn More
Sino Biological Recombinant Human c-MET/HGFR Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10692H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human c-Met (NP_000236.2) (Met1-Thr932) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5418