Learn More
Sino Biological Recombinant Human EpCAM/TROP1 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10694H08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Lys265) of human EpCAM (NP_002345.1) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-161-5427