Learn More
Sino Biological Recombinant Human Neuropilin-2/NRP2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10695H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Tyr 855) ofhuman Neurophilin2 isoform 2 (NP_003863.2) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5431