Learn More
Sino Biological Recombinant Human B7-1/CD80 Protein
Supplier:  Sino Biological 10698HCCH100
Encompass_Preferred
The mature form of human B7-1 (NP_005182.1) extracellular domain (Met 1-Asn 242) with five amini acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-5444