Learn More
Sino Biological Recombinant Human TIE-2 Protein (ECD, His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 10700H03H200
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Lys745) of human Tie2 (AAA61139.1) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5453