Learn More
Sino Biological Recombinant Human GM-CSF Receptor alpha Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10701H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gly 320) of human GM-CSFRα (NP_006131.2) pro-protein was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-5459