Learn More
Sino Biological Recombinant Human GM-CSF Receptor alpha Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10701H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gly 320) of human GM-CSFRα (P15509-1) was expressed with the C-terminal fused polyhistidine tag.
Catalog No. 50-161-5461