Learn More
Sino Biological Beta-amyloid 42 / Beta-APP42 Protein, Human, Recombinant (His & GST Tag)
Supplier:  Sino Biological 10703H20E2100
Encompass_Preferred
A DNA sequence encoding the amino acids (Asp672-Ala713) of human Amyloid beta A4 protein (APP770) (P05067-1), corresponding to the Beta-amyloid protein 42, was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-5472