Learn More
Sino Biological Recombinant Human IFN-beta Protein (hFc Tag)
Supplier:  Sino Biological 10704H02H100
Encompass_Preferred
A DNA sequence encoding the human IFNβ (NP_002167.1) (Met 1-Asn 187) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5478