Learn More
NEST SCIENTIFIC INC 1070UL ROBOTIC TIPS 30/CS
Catalog No. NC3650185
Encompass_Preferred
1070uL automation tips for use with Beckman liquid handlers. Sterile and packed in 12X8 SBS/ANSI footprint racks. Lot tested in house on Beckman i7 to ensure proper fit and CV values. Molded in ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility. 2880 tips per case.
Catalog No. NC3650185 Supplier NEST SCIENTIFIC INC Supplier No. 317011
1070uL automation tips for use with Beckman liquid handlers. Sterile and packed in 12X8 SBS/ANSI footprint racks. Lot tested in house on Beckman i7 to ensure proper fit and CV values. Molded in ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility. 2880 tips per case.