Learn More
Sino Biological Recombinant Human CLK3 Protein (GST Tag)
Supplier:  Sino Biological 10716H09B50
Encompass_Preferred
A DNA sequence encoding the human CLK3 isoform b (NP_003983.2) (Met 1-Arg 490) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5497