Learn More
Sino Biological Recombinant Human TRIB2 Protein (His & GST Tag)
Supplier:  Sino Biological 10725H19B50
Encompass_Preferred
A DNA sequence encoding the human TRIB2 (NP_067675.1) (Met 1-Asn 343) was fused with the GST tag at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5502