Learn More
Sino Biological Recombinant Human DDR1 Protein (aa 444-913, His & GST Tag)
Supplier:  Sino Biological 10730H20B150
Encompass_Preferred
A DNA sequence encoding the human DDR1 (Q08345-1) (Arg444-Val913) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-5515