Learn More
Sino Biological Recombinant Human IRAK4 Protein (His Tag)
Catalog No. 501615521
Encompass_Preferred
A DNA sequence encoding the full length of human IRAK4 (Q9NWZ3-1) (Met 1-Ser 460) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-5521 Supplier Sino Biological Supplier No. 10735H07B50
A DNA sequence encoding the full length of human IRAK4 (Q9NWZ3-1) (Met 1-Ser 460) was expressed with a polyhistidine tag at the N-terminus.