Learn More
Sino Biological Recombinant Human B7-H4 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10738H08H100
Encompass_Preferred
A DNA sequence encoding the human VTCN1 (Q7Z7D3-1) (Phe29-Ala258) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5526