Learn More
Sino Biological Recombinant Human CDK1/CDC2 Protein (GST Tag)
Supplier:  Sino Biological 10739H09B50
Encompass_Preferred
A DNA sequence encoding the human CDC2 isoform 1 (NP_001777.1) (Met 1-Met 297) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5528