Learn More
Sino Biological Recombinant Human CD42c Protein (His Tag)
Supplier:  Sino Biological 10742H08H100
Encompass_Preferred
A DNA sequence encoding the human GPIb, β polypeptide extracellular domain (NP_000398.1) (Met 1-Cys 147) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5531