Learn More
Sino Biological Recombinant Human JNK2 Protein (His Tag)
Supplier:  Sino Biological 10745H08B50
Encompass_Preferred
A DNA sequence encoding the full length of human MAPK9 (NP_002743.3) (Met 1-Arg 424) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5534