Learn More
Sino Biological Recombinant Human P38 Delta/MAPK13 Protein (GST Tag)
Supplier:  Sino Biological 10747H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human MAPK13 (O15264) (Met 1-Leu 365) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5536