Learn More
Sino Biological Recombinant Human VAPB Protein (His Tag)
Supplier:  Sino Biological 10754H08E100
Encompass_Preferred
A DNA sequence encoding the human VAPB (O95292-1) N-terminal fragment (Met 1-Pro 132) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5543