Learn More
Sino Biological Recombinant Human ZIP Kinase/DAPK3 Protein (GST Tag)
Supplier:  Sino Biological 10757H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human DAPK3 (NP_001339.1) (Met 1-Arg 454) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5548