Learn More
Sino Biological Recombinant Human MICB Protein (His & hFc Tag)
Supplier:  Sino Biological 10759H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human MICB (NP_005922.2) (Met 1-Gly 298) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5551