Learn More
Sino Biological Recombinant Human MICB Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10759H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human MICB (NP_005922.2) (Met 1-Gly 298) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5553