Learn More
Sino Biological Recombinant Human EphB2 Protein (His & hFc Tag)
Supplier:  Sino Biological 10762H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Leu 543) of human EphB2 (NP_059145.2) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5555