Learn More
Sino Biological Recombinant Human AKT1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10763H08B50
Encompass_Preferred
A DNA sequence encoding the human AKT1 (NP_001014431.1) (Met 1-Ala 480) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5564