Learn More
Sino Biological Recombinant Human AKT3 Protein (GST Tag)
Supplier:  Sino Biological 10765H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human PKBG (NP_005456.1) (Met 1-Glu 479) was expressed with the GST tag at the N-terminus.
Catalog No. 50-161-5568