Learn More
Sino Biological Recombinant Human IP-10/CXCL10 Protein
Supplier:  Sino Biological 10768HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human CXCL10 (NP_001556.2) (Val22-Pro98) was expressed, with a N-terminal Met.
Catalog No. 50-161-5571