Learn More
Sino Biological Recombinant Human ASGR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10773H07H100
Encompass_Preferred
A DNA sequence encoding the human ASGR1 isoform 1 (P07306) (Gln 62-Leu 291) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-5577