Learn More
Sino Biological Recombinant Human CD40/TNFRSF5 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10774H02H100
Encompass_Preferred
A DNA sequence encoding the human CD40 (NP_001241.1) (Met1-Arg193) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5579