Learn More
Sino Biological Recombinant Human TGF-beta RIII/TGFBR3 Protein (His Tag)
Supplier:  Sino Biological 10778H08B100
Encompass_Preferred
A DNA sequence encoding the human TGFBR3 (Q03167-1) (Met 1-Gly781) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5590