Learn More
Sino Biological Recombinant Human CD70/CD27 Ligand Protein (hFc Tag)
Catalog No. 501615592
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD70 (NP_001243.1) (Gln 39-Pro 193) was fused with the Fc region of human IgG1 at the N-terminus via a polypeptide linker.
Catalog No. 50-161-5592 Supplier Sino Biological Supplier No. 10780H01H100
Add to Cart
Add to Cart
A DNA sequence encoding the extracellular domain of human CD70 (NP_001243.1) (Gln 39-Pro 193) was fused with the Fc region of human IgG1 at the N-terminus via a polypeptide linker.