Learn More
Sino Biological Recombinant Human BLK Protein (GST Tag)
Supplier:  Sino Biological 10782H09B50
Encompass_Preferred
A DNA sequence encoding the human BLK (NP_001706.2) (Met 1-Pro 505) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5604