Learn More
Sino Biological Recombinant Human CD5L Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10791H08H100
Encompass_Preferred
A DNA sequence encoding the human CD5L (NP_005885.1) (Met 1-Gly 347) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5607