Learn More
Sino Biological Recombinant Human JNK1 Protein (GST Tag)
Supplier:  Sino Biological 10795H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human MAPK8 isoform JNK1 alpha 2 (NP_620637.1) (Met 1-Arg 427) was expressed with the GST tag at the N-terminus.
Catalog No. 50-161-5612