Learn More
Sino Biological Recombinant Human CD48 Protein (His Tag)
Supplier:  Sino Biological 10797H08H100
Encompass_Preferred
A DNA sequence encoding the human CD48 (NP_001769.2) (Met 1-Ser 220) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5615