Learn More
Sino Biological Recombinant Human CXADR/CAR Protein (His & hFc Tag)
Supplier:  Sino Biological 10799H03H100
Encompass_Preferred
A DNA sequence encoding the human CXADR (NP_001329.1) extracellular domain (Met 1-Gly 237) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5617