Learn More
Ace Glass, Inc. -108 FETFE O-RING 12/PK -108 FETFE O-RING 12/PK
SDP
Catalog No. 509558048
Encompass_Preferred
Size 108 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-955-8048 Supplier Ace Glass, Inc. Supplier No. 7855711
Size 108 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.