Learn More
Sino Biological Recombinant Human MAP3K8 Protein (GST Tag)
Supplier:  Sino Biological 10800H09B50
Encompass_Preferred
A DNA sequence encoding the human MAP3K8 isoform 1 (P41279-1) (Met 30-Arg 397) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5622