Learn More
Sino Biological Recombinant Human TGF beta 1 Protein (Latent, His Tag)
Supplier:  Sino Biological 10804H08H50
Encompass_Preferred
A DNA sequence encoding the full length of human TGFβ1 (NP_000651.3) (Met 1-Ser 390) was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-5627