Learn More
Sino Biological Recombinant Human, Rhesus, Cynomolgus, Canine TGF beta 1 Protein
Supplier:  Sino Biological 10804HNAC100
Encompass_Preferred
A DNA sequence encoding the active form of human / rhesus / cynomolgus / canine TGFβ1 (NP_000651.3) (Ala 279-Ser 390) was expressed and purified. Human, Rhesus, cynomolgus and Canine TGFβ1 sequences are identical.
Catalog No. 50-161-5629