Learn More
Sino Biological Recombinant Human VEGFR3/FLT4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10806H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ile 776) of human VEGFR3 (NP_002011.2) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5641