Learn More
Sino Biological Recombinant Human Uteroglobin Protein (His Tag)
Supplier:  Sino Biological 10808H08H100
Encompass_Preferred
A DNA sequence encoding the human SCGB1A1 (P11684) (Met 1-Asn 91) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-5643