Learn More
Sino Biological Recombinant Human CD21 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10811H08H100
Encompass_Preferred
A DNA sequence encoding the human CD21 isoform A (P20023-1) extracellular domain (Met 1-Arg 971) was expressed, with a carboxy-terminal polyhistidine tag.
Catalog No. 50-161-5647