Learn More
Sino Biological Recombinant Human CD38 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10818H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD38 (NP_001766.2) (Val 43-Ile 300) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5657