Learn More
Sino Biological Recombinant Human CD38 Protein (His & FLAG Tag), HPLC-verified
Supplier:  Sino Biological 10818H32H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD38 (NP_001766.2) (Val 43 – Ile 300) was fused with a polyhistidine tag and a flag tag at the N-terminus.
Catalog No. 50-161-5661