Learn More
Sino Biological Recombinant Human CEACAM-1/CD66a Protein (ECD, His & hFc Tag)
Supplier:  Sino Biological 10822H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CEACAM1 (NP_001020083.1) (Met 1-Gly 428) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5666