Learn More
Sino Biological Recombinant Human CEACAM-6/CD66c Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10823H08H100
Encompass_Preferred
A DNA sequence encoding the human CEACAM6 (NP_002474.4,with natural variants V239G) (Met1-Gly320) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5670